Mueller Hinton Agar II, BD
Supplier: BD
Certificates
About this item
Mueller-Hinton agar is a microbiological growth medium that is commonly used in antibiotic susceptibility testing.
- Non-selective, non-differential medium
- Contains starch
- Loose agar for better diffusion of antibiotics
- Truer zone of inhibition
Agar is also used to isolate and maintain Neisseria and Moraxella species.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.