Germ Tube Cryo™ for Candida Albicans, Hardy Diagnostics
10324-568
Product available on GSA Advantage®
:
- Description:Germ Tube Cryo™ for Candida Albicans
- Size:0.5 mL
- Cat. No.:10324-568
- Supplier no.:Z217
A ready-to-use germ tube solution made up of stabilized animal serum.
- Comes pre-measured in vials
- No mixing, measuring, or aliquoting is required
- Eliminates exposure to human serum
A ready-to-use germ tube solution made up of stabilized animal serum, comes pre-measured in vials. No mixing, measuring, or aliquoting is required! Eliminates exposure to human serum. 0.5ml in each vial. Germ tubes develop on C. albicans within one hour. Serum formula is enhanced for more rapid and luxuriant production of germ tubes.