Anti-KIR3DL2 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 539304]
103238-728
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:KIR3DL2/CD158k
- Antigen Symbol:KIR3DL2
- Clonality:Monoclonal
- Clone:539304
- Conjugation:PE (Phycoerythrin)
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:25 Tests
- Storage Buffer:Supplied in a saline solution containing BSA and Sodium Azide.
- Storage Temperature:Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
- Immunogen:BaF3 mouse pro-B cell line transfected with human KIR3DL2/CD158k Accession # P43630
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:103238-728
- Supplier no.:FAB2878P-025
Type: Primary Antibodies
Antigen: KIR3DL2
Clonality: Monoclonal
Clone: 539304
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human