Rapid MGP Media for Enterococcus
10324-570
Product available on GSA Advantage®
:
- Description:Rapid MGP Media for Enterococcus
- Size:1 mL
- Cat. No.:10324-570
- Supplier no.:Z225
A five hour test for the acidification of MGP.
- Used to rule out non-pathogenic E. casseliflavus and E. gallinarum
- aid in the differentiation of clinically significant Enterococcus faecalis and fecium
A five hour test for the acidification of MGP to aid in the differentiation of clinically significant Enterococcus faecalis and fecium. Used to rule out non-pathogenic E. casseliflavus and E. gallinarum.