MycoVue™, Slide Culture System, Potato Flake Agar, Ready-To-Use, Hardy Diagnostics
Catalog # 10324-474
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:MycoVue™ Slide Culture System, Potato Flake Agar, Ready-To-Use
- Size:5 trays per box
- Cat. No.:10324-474
- Supplier no.:MV1
Specifications
About this item
A ready-to-use slide culture system for the cultivation of fungi.
- Simplify your fungal slide cultures while saving money
- Culture can be incubated for long periods of time
- Easy observation without disturbing the fungal structures
A ready-to-use slide culture system for the cultivation of fungi. Simplify your fungal slide cultures while saving money, reducing frustration, and eliminating preparation time. Desiccation is prevented with a built in humidity chamber and the culture can be incubated for long periods of time. Device fits conveniently on the microscope stage for easy observation without disturbing the fungal structures.