Order Entry
Puerto Rico
ContactUsLinkComponent
Thioglycollate with Supplements, Filtered , Hardy Diagnostics
Thioglycollate with Supplements, Filtered , Hardy Diagnostics
Catalog # 10324-458
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Thioglycollate with Supplements, Filtered , Hardy Diagnostics
Catalog # 10324-458
Supplier:  Hardy Diagnostics
Supplier Number:  K23
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Thioglycollate with Supplements, Filtered
  • Size:
    10 mL
  • Cat. No.:
    10324-458
  • Format:
    16×125 mm tube
  • Supplier no.:
    K23

Specifications

About this item

For the cultivation of fastidious anaerobes.

  • Filtered to remove all dead bacteria and debris
  • Intended for human diagnostic use
  • Filtering ensures medium becomes clear, clean, and free from dead bacteria

(Thioglycollate without Indicator, with Hemin, Vitamin K, and Calcium Carbonate Chip as a buffer) For the cultivation of fastidious anaerobes

Intended for human diagnostic use.

The last thing you want to see on your Gram Stain of sterile fluids is dead bacteria! Not from your specimen, but from your culture medium broth…

Did you know that all powdered culture media contains bacteria that, although killed during the sterilization process, may show up as dead bacteria on your Gram stain? This is a possible cause of false positive reports. By filtering Thioglycollate before sterilization, the medium becomes clear, clean, and free from dead bacteria! Avoid false positives, and insist that your Thioglycollate be filtered.