Rose Agar for Gram Positives, Hardy Diagnostics
Catalog # 10324-338
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:Rose Agar for Gram Positives
- Size:Pack of 10
- Cat. No.:10324-338
- Format:15×100 mm plate
- Supplier no.:A66
Specifications
About this item
For the selective isolation of gram positive bacteria.
- CAN and PEA combination
- Selective isolation of gram positive bacteria
(CNA and PEA Combination) For the selective isolation of gram positive bacteria.