ContactUsLinkComponent
Q-Slide™ AFB, Prepared Control Slide for QC of AFB Stain, Hardy Diagnostics
Q-Slide™ AFB, Prepared Control Slide for QC of AFB Stain, Hardy Diagnostics
Catalog # 10119-786
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
undefined
Q-Slide™ AFB, Prepared Control Slide for QC of AFB Stain, Hardy Diagnostics
Catalog # 10119-786
Supplier:  Hardy Diagnostics
Supplier Number:  Z301
Product available on GSA Advantage®
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Q-slide™ afb, prepared control slides for qc of afb stain
  • Cat. No.:
    10119-786
  • Supplier no.:
    Z301
Specifications
About this item
Hardy’s new Q-Slide™ is a quick and effective way to ensure the quality of reagents used as well as the proficiency of staining technique by laboratory personnel.
  • Convenient, ready-to-stain control slide
  • Process four patient specimens at the same time
  • Q-Slide™ AFB - For your Acid Fast Bacillus Stain QC
  • Q-Slide™ Gram - For your Gram Stain QC
A convenient ready-to-stain control slide that will test both your stain solutions and your staining techniques
Q-Slide™ AFB - For your Acid Fast Bacillus Stain QC; either Kinyoun or Ziehl Neelson. Contains Mycobacterium gordonae as a positive control and a Corynebacterium spp. as a negative control.
Q-Slide™ Gram - For your Gram Stain QC. Contains E.coli as a gram-negative control, and a Staphylococcus aureus as a gram-positive control.
Positive and negative stain control organisms are affixed to the circles in each Q-Slide™. Up to four patient specimens (in the squares) may be processed at the same time as the QC organisms.