Attest™ Rapid Readout Biological Indicators and Test Packs for EO, 3M™
Supplier: Healthcare Products
Certificates
About this item
Biological indicators and test packs monitor the effectiveness of the EO sterilization process.
- Biological Indicator results in four hours
- Every load monitoring and quarantining can eliminate recalls
- Malfunctioning sterilizers can be repaired and put back into service quickly
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.