Order Entry
Puerto Rico
ContactUsLinkComponent
 
HardyDisks™ Lysostaphin
Catalog # 10136-070
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
HardyDisks™ Lysostaphin
Catalog # 10136-070
Supplier:  Hardy Diagnostics
Supplier Number:  Z112
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Description:
    Lysostaphin
  • Size:
    50 Disks
  • Cat. No.:
    10136-070
  • Application:
    Clinical
  • Supplier no.:
    Z112

Specifications

About this item

Differentiates between Micrococcus and Staphylococcus.

Used to rapidly differentiate Staphylococcus spp. and Micrococcus spp. based on lysostaphin resistance. Make a suspension of the bacteria, add the disk, look for a clearing of turbidity if Staph is present.