ContactUsLinkComponent
Arizant Bair Hugger™ Spinal Underbody Warming Blanket, 3M™
Arizant Bair Hugger™ Spinal Underbody Warming Blanket, 3M™
  75787-610
Arizant Bair Hugger™ Spinal Underbody Warming Blanket, 3M™
  75787-610
 :  MMM 57501
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Model 575 Spinal Underbody Warming Blanket, with 61 x 122 cm (24 x 48") Drape
  • Size:
    188 x 91.4 cm (74 x 36")
  • Environmentally Preferable:
  • Cat. No.:
    75787-610
  • Supplier no.:
    MMM57501
This advanced forced-air warming blanket works with the open frame of the spinal surgery cradle and does not interfere with the adjustment of support pads.
  • Integrated tie strips secure the blanket to the table frame
  • Blanket design allows it to draw up near the patient when inflated
  • Soft, radiolucent materials accommodate imaging requirements
  • One large clear plastic drape (included) helps retain warmth air around the patient's head
  • Compatible with foot boards, leg sling, Wilson frame, or head support modalities
Perforations on the side of the blanket allow the table frame to pass through the blanket. Removable section allows visualization of the patient's face throughout the procedure.