Order Entry
Puerto Rico
ContactUsLinkComponent
Enucleation Scissors, OR Grade, Sklar
Enucleation Scissors, OR Grade, Sklar
Catalog # 10197-846
Supplier:  Sklar
undefined
Enucleation Scissors, OR Grade, Sklar
Catalog # 10197-846
Supplier:  Sklar
Supplier Number:  64-1150
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Length:
    127 mm (5")
  • Material:
    Stainless steel
  • Curvature:
    Slightly Curved
  • Cat. No.:
    10197-846
  • Supplier no.:
    64-1150
  • Blade type:
    Blunt/Blunt

Specifications

About this item

Enucleation Scissors are an ophthalmic instrument used primarily in procedures dealing with removal of the globe due to tumor or trauma

These finger ring scissors have thick blades that allow the surgeon to manuever around the globe and reach the optic nerve in order to sever it. The edges are smooth and the tips are blunt.