Xylene (mixture of isomers) ≥99.00%
Catalog # 71009-384
Supplier: KMG
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Poly bottle
- Size:1 gal.
- Cat. No.:71009-384
- Supplier no.:464-200554
Specifications
Specification Test Results
- Assay (Xylene):>99.00
- Assay (H2O):0,20%
- Color:10
- Acidity:0.300µeq/g
- Residue After Evaporation:5000 ppb
- Chloride:3000 ppb
- Phosphate:1000 ppb
- Sulfur Compounds:30 ppb
- Infared:Conform
- Appearance:Conform
- Sustance Darkening (by H2SO4):Conform
- Aluminum:100 ppb
- Antimony:2 ppb
- Arsenic:10 ppb
- Boron:100 ppb
- Calcium:100 ppb
- Chromium:100 ppb
- Copper:10 ppb
- Gold:100 ppb
- Iron:100 ppb
- Lead:100 ppb
- Magnesium:100 ppb
- Manganese:100 ppb
- Nickel:10 ppb
- Potassium:100 ppb
- Sodium:100 ppb
- Tin:100 ppb
- Titanium:100 ppb
- Zinc:100 ppb
- 1.0µ Particle Count:10 par/ml
Specification Test Results
About this item
:
:106.17 g/mol
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.86…0.88 g/cm³ (20 °C)
Flash Pt:25 °C
UN:1307
ADR:3,III
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264