Order Entry
Puerto Rico
ContactUsLinkComponent
QBC® STAR™ Dry Hematology Analyzer, Drucker Diagnostics
QBC® STAR™ Dry Hematology Analyzer, Drucker Diagnostics
Catalog # 10755-958
Supplier:  Drucker
undefined
QBC® STAR™ Dry Hematology Analyzer, Drucker Diagnostics
Catalog # 10755-958
Supplier:  Drucker
Supplier Number:  429001

Specifications

  • Sample Volume:
    70 µl
  • Description:
    QBC® STAR™ Dry Hematology, 100 - 240 VAC
  • Dimensions:
    40.6W×41.4D×41.4H (16×16.3×16.3")
  • Operating Temperature:
    16...32 °C
  • Weight:
    8.6 kg (19 lbs.)
  • Cat. No.:
    10755-958
  • Supplier no.:
    429001

Specifications

About this item

The QBC STAR™ dry hematology system is the gold standard for performing CBC testing in a variety of settings, including office labs and even remote locations. It employs innovative dry hematology technology, meaning that all of the reagents needed to perform a CBC test with it are contained in the QBC STAR™ tube. After filling the tube with the blood samples, the clinician simply places it in the specially designed QBC STAR™ system and obtains the test results after the analysis is complete. This unique approach provides multiple benefits for in-office CBC testing.

  • Fully automatic
  • Simple, single button operation
  • No liquid reagents
  • Nine parameter, two part differential CBC
  • For capillary and venous blood
  • Accurate reads without the calibration
  • Built-in printer prints results on 58 mm thermal recorder paper
  • 3 external USB ports

The testing items include hematocrit (Hct) (%), hemoglobin (Hgb) (g/dl).

One-year warranty