TransIT-PRO® Transfection Reagent Kit, Mirus Bio
Supplier: Mirus Bio
Certificates
About this item
Ideal for biotherapeutic protein production in suspension CHO and 293 cells.
- High protein yield
- Compatible with multiple CHO formulations
- Simple workflow
Reach critical stages faster. Decrease time to produce usable protein by maximizing target protein yields through transient transfection. The TransIT-PRO® Transfection Reagent & Kit use animal origin free components designed for high and reproducible protein yield in suspension CHO and 293 derived cells. Since it is compatible with varied media formulations, the same media can be used for both transient and stable expression. Use with CHOgro® Expression Media for highest yield in suspension CHO cells. TransIT-PRO® outperforms linear PEI in protein yield, while providing a cost-effective alternative to FreeStyle™ MAX.
Specifications
- Bioproduction:Yes
- Cell Specific:293, CHO
- DNA:Yes
- Virus production:Yes