TransIT®-mRNA Transfection Kit, Mirus Bio
A high-efficiency, low-toxicity transfection reagent for large RNA and CRISPR guide RNA.
- Low cellular toxicity
- High-efficiency delivery
- Serum compatible
- Deliver various sizes of RNA
Use these kits to maintain cell density and reduce experimental biases, as well as perform transfection in the presence of serum, eliminating the need for a medium change and helping to maintain cellular health.
Each kit is supplied with the TransIT®-mRNA Transfection Reagent and the mRNA Boost Reagent.
- CRISPR/Cas 9:Yes
- mRNA:Yes
- Virus production:Yes