Order Entry
Puerto Rico
ContactUsLinkComponent
Zinc Standard, Titrisol®, MilliporeSigma
Zinc Standard, Titrisol®, MilliporeSigma
Catalog # EM1.09953.0001
Supplier:  MilliporeSigma
CAS Number:  
Zinc Standard, Titrisol®, MilliporeSigma
Catalog # EM1.09953.0001
Supplier:  MilliporeSigma
Supplier Number:  1.09953.0001
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    H₂O/tr. HCl
  • Pack type:
    Plastic Ampule
  • Size:
    1 Ampule
  • Cat. No.:
    EM1.09953.0001
  • pH:
    2.0 (H₂O, 20°C)
  • Element:
    Zinc
  • Supplier no.:
    1.09953.0001

Specifications

About this item