Air-Mate™ Powered Air Purifying Respirator Components, 3M™
: Avantor
Powered Air Purifying Respirators (PAPR) are motorized systems powered by a battery pack which draws air through a filter or cartridge to provide respiratory protection to the wearer.
- Loose-fitting headgear options provide respiratory protection without the need for a tight faceseal or fit testing
- Battery-powered blower pulls air through the filter with no additional breathing resistance to the wearer
- Battery and filter internal to PAPR; no power cords required
PAPR system components are sold individually.
The blower unit can be mounted on the waist, face, or head, depending on the system selected.