4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) substrate 5 mg, tablets, Reagent Grade
Supplier: VWR
Certificates
About this item
CAS Number:333338-18-4
:4264-83-9
Formula:C₆H₄NNa₂O₆P·6H₂O
MDL Number:MFCD00066288
:371.144 g/mol
Storage Temperature:Refrigerator
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Abs.@309nm (1 tab/250mL Water):0.48 - 0.58
- Background Color:<= 0.14
- Mean Tablet Weight:170 mg - 180 mg