Anti-GRIN2B Mouse Monoclonal Antibody [clone: NR2B]
10168-832
: Genetex
- Antibody Type:Primary
- Antigen Name:Glutamate Receptor, Ionotropic, N-Methyl D-Aspartate 2B
- Antigen Symbol:GRIN2B
- Clonality:Monoclonal
- Clone:NR2B
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:2904
- Antigen Synonyms:hNR3|GluN2B|NR2B|EIEE27|MRD6|NMDAR2B
- Storage Buffer:PBS containing 0.05 % sodium azide and 1 mg/ml BSA
- Storage Temperature:–20 °C, avoid freeze/thaw cycles
- Concentration:1 mg/ml
- Immunogen:Recombinant protein: NR2B subunit of amino acid residues 934-1457.
- Purification:Protein A
- Cat. No.:10168-832
- Supplier no.:GTX80691
Mouse monoclonal antibody [NR2B] to NMDA Receptor 2B
Type: Primary
Antigen: GRIN2B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat