To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Influenza A Virus H1N1 Hemagglutinin (HA)
- Antigen Symbol:N1 HA
- Clonality:Monoclonal
- Clone:GT223
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Influenza A
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Storage Buffer:PBS
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant fragment corresponding to C-terminal region of Influenza A virus Hemagglutinin (HA) (A/WSN/1933(H1N1)). The exact sequence is proprietary.
- Purification:Affinity purified by Protein G.
- Cat. No.:10167-476
- Supplier no.:GTX629750
Specifications
About this item
Mouse Monoclonal antibody [GT223] to Influenza A Virus H1N1 Hemagglutinin (HA) (Influenza A H1N1 Hemagglutinin(HA))
Type: Primary
Antigen: N1 HA
Clonality: Monoclonal
Clone: GT223
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Influenza A virus