Anti-His tag Antibody [GT359] Mouse Monoclonal Antibody [clone: GT359] (HRP (Horseradish Peroxidase))
Catalog # 10168-050
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Symbol:His tag antibody [GT359]
- Clonality:Monoclonal
- Clone:GT359
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- Isotype:IgG3
- Western Blot:Yes
- Size:100 μl
- Storage Buffer:0.05M Tris, 0.15M NaCl (pH7.4). 0.025% ProClin 300 was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:0.16 mg/ml
- Purification:Affinity purified by Protein G.
- Cat. No.:10168-050
- Supplier no.:GTX628914-01
Specifications
About this item
HRP-conjugated Mouse Monoclonal antibody [GT359] to 6X His tag
Type: Primary
Antigen: His
Clonality: Monoclonal
Clone: GT359
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: