Anti-MYC Mouse Monoclonal Antibody [clone: GT168]
Catalog # 89497-590
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:V-Myc Avian Myelocytomatosis Viral Oncogene Homolog
- Antigen Symbol:MYC
- Clonality:Monoclonal
- Clone:GT168
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:200 μl
- Gene ID:4609
- Antigen Synonyms:MRTL|bHLHe39|c-Myc|MYCC
- Storage Buffer:PBS
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Purification:Affinity purified by Protein G.
- Cat. No.:89497-590
- Supplier no.:GTX628459
Specifications
About this item
Mouse Monoclonal antibody [GT168] to c-Myc (v-myc myelocytomatosis viral oncogene homolog (avian))
Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human