To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Histone Cluster 1, H3A
- Antigen Symbol:HIST1H3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:50 μl
- Environmentally Preferable:
- ChIP:Yes
- Gene ID:8350
- Antigen Synonyms:H3FA|H3/A
- Storage Buffer:Whole antiserum from rabbit containing 0.05% azide.
- Storage Temperature:Store at –20 °C; for long storage, store at –80 °C. Avoid multiple freeze-thaw cycles.
- Purification:Whole antiserum from rabbit containing 0.05% azide.
- Cat. No.:10165-570
- Supplier no.:GTX60327
Specifications
About this item
Rabbit polyclonal antibody to Histone H3K9me3S10ph (trimethyl Lys9, phospho Ser10)
Type: Primary
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human