Anti-AOC3 Mouse Monoclonal Antibody [clone: 174-5]
Catalog # 10794-502
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Amine Oxidase, Copper Containing 3
- Antigen Symbol:AOC3
- Clonality:Monoclonal
- Clone:174-5
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:50 μg
- Gene ID:8639
- Antigen Synonyms:VAP-1.|Vascular adhesion protein 1|Semicarbazide-sensitive amine oxidase|Membrane primary amine oxidase|Membrane copper amine oxidase|Semicarbazide sensitive amine oxidase|Vascular Adhesion Protein 1|HPAO|SSAO|AOC3|VP97|AOC3_HUMAN|Amine Oxidase|Copper Containing 3 (vascular adhesion protein 1)|Copper amine oxidase
- Storage Buffer:PBS, containing 0.1% bovine serum albumin.
- Storage Temperature:Product should be stored at 4 °C. Under recommended storage conditions, product is stable for at least one year. The exact expiry date is indicated on the label.
- Immunogen:Purified vessels from human peripheral lymph nodes (ref 1)
- Cat. No.:10794-502
Specifications
About this item
Mouse Monoclonal antibody [174-5] to VAP1
Type: Primary
Antigen: AOC3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human