Anti-CD163 Mouse Monoclonal Antibody [clone: RM3/1]
10793-218
: Genetex
- Antibody Type:Primary
- Antigen Name:Cd163 Molecule
- Antigen Symbol:CD163
- Clonality:Monoclonal
- Clone:RM3/1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Size:50 μg
- Gene ID:9332
- Antigen Synonyms:M130|MM130
- Storage Buffer:PBS, containing 0.02% sodium azide and 0.1% bovine serum albumin.
- Storage Temperature:Product should be stored at 4 °C. Under recommended storage conditions, product is stable for one year.
- Immunogen:Human monocytes (epitope recognized SRCR domain 9)
- Cat. No.:10793-218
Mouse Monoclonal antibody [RM3/1] to CD163
Type: Primary
Antigen: CD163
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse