Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-EMP1 Rabbit Polyclonal Antibody
Anti-EMP1 Rabbit Polyclonal Antibody
Catalog # 10164-606
Supplier:  Genetex
Anti-EMP1 Rabbit Polyclonal Antibody
Catalog # 10164-606
Supplier:  Genetex
Supplier Number:  GTX50979

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Epithelial Membrane Protein 1
  • Antigen Symbol:
    EMP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Size:
    100 μg
  • Gene ID:
    2012
  • Antigen Synonyms:
    EMP-1|CL-20|TMP
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and less than 0.09% sodium azide.
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    KLH conjugated synthetic peptide derived from mouse Emp1
  • Purification:
    Purified by Protein A and peptide affinity chromatography.
  • Cat. No.:
    10164-606
  • Supplier no.:
    GTX50979

Specifications

About this item

Rabbit Polyclonal antibody to Emp1

Type: Primary
Antigen: EMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat