Anti-E2f7 Rabbit Polyclonal Antibody
50 μg
Catalog # 10794-650
Supplier: Genetex
: GTX48989
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit polyclonal antibody to E2F7
Type: Primary
Antigen: E2f7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Dog, Pig, Rat, Horse, Rabbit, Bovine
Specifications
- Size:50 μg
- Clonality:Polyclonal
- Purification:Antigen affinity purification
- Storage Buffer:PBS contains 0.09% (w/v) sodium azide and 2% sucrose
- Isotype:IgG
- Host:Rabbit
- Cat. No.:10794-650
- Reactivity:Pig|Rabbit|Horse|Human|Bovine|Dog|Rat|Mouse
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:E2f7
- Gene ID:52679
- Immunogen:Synthetic peptide located within the following region: MEVNCLTLKDLISPRQTRLDFAIEDAENAQKENIFVDRSRMTPKTPMKNE
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Name:E2f7
- Storage Temperature:For longer periods of storage, store at -20C. Avoid repeat freeze-thaw cycles.



