Anti-AOC1 Rabbit Polyclonal Antibody
Catalog # 10792-226
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Amine Oxidase, Copper Containing 1
- Antigen Symbol:AOC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:26
- Antigen Synonyms:ABP1|DAO|KAO|DAO1|ABP
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
- Storage Temperature:Store at –20 °C (regular) or –80 °C (long term). Avoid freeze / thaw cycles.
- Immunogen:Recombinant protein of human AOC1
- Purification:Antigen affinity purification
- Cat. No.:10792-226
Specifications
About this item
Rabbit polyclonal antibody to KAO
Type: Primary
Antigen: AOC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat