Anti-XRCC3 Rabbit Polyclonal Antibody
Catalog # 10168-390
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:X-Ray Repair Complementing Defective Repair In Chinese Hamster Cells 3
- Antigen Symbol:XRCC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:7517
- Antigen Synonyms:CMM6
- Storage Buffer:PBS with 0.05% sodium azide
- Storage Temperature:Upon receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:5.3 mg/ml
- Immunogen:Synthetic partial peptide of Human XRCC3.
- Purification:Immunogen affinity purified
- Cat. No.:10168-390
- Supplier no.:GTX26494
Specifications
About this item
Rabbit polyclonal to XRCC3
Type: Primary
Antigen: XRCC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human