Anti-SLCO1B1 Mouse Monoclonal Antibody [clone: ESL]
10167-910
: Genetex
- Antibody Type:Primary
- Antigen Name:Solute carrier organic anion transporter Family member 1B1
- Antigen Symbol:SLCO1B1
- Clonality:Monoclonal
- Clone:ESL
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Size:100 μl
- Gene ID:10599
- Antigen Synonyms:OATP-C|HBLRR|LST-1|OATP1B1|OATP2|SLC21A6|OATPC|LST1
- Storage Buffer:tissue culture supernatant containing 0.09 % sodium azide
- Storage Temperature:–20 °C, avoid freeze/thaw cycles
- Immunogen:Synthetic peptide corresponding to residues E(671) S L N K N K H F V P S A G A D S E T H C(691) of human OATP2.
- Cat. No.:10167-910
- Supplier no.:GTX15441
Mouse monoclonal antibody [ESL] to OATP2
Type: Primary
Antigen: SLCO1B1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human