Anti-PRPF31 Rabbit Polyclonal Antibody
Catalog # 10166-458
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Pre-mRNA Processing Factor 31
- Antigen Symbol:PRPF31
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:26121
- Antigen Synonyms:RP11|SNRNP61|NY-BR-99|PRP31
- Storage Buffer:1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human PRPF31. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:10166-458
- Supplier no.:GTX117081
Specifications
About this item
Rabbit Polyclonal antibody to PRPF31 (pre-mRNA processing factor 31)
Type: Primary
Antigen: PRPF31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat