Disposable Samplers, MultiDispo, Burkle
Supplier: BURKLE INC
Certificates
About this item
Disposable zone samplers for taking samples from multiple layers of the sample material.
- HDPE
- Clean room manufactured
- Optionally sterilised by gamma rays
- Easy emptying through the opening in the handle (Ø 14 mm)
Suitable for free-flowing powders and small-grain granulates. The representative all-layer sample can be emptied directly through the open handle into a sample container.
Specifications
- Catalog No:
- 10821-281
- 10821-261
- 10821-279
- 10821-277
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- ISO 14001 Certificate
- ISO 14001 Certificate
- ISO 14001 Certificate
- ISO 14001 Certificate