RNase Erase™ removes RNase contamination from glass, plastic, pipettes, and equipment.
- Removes RNase contamination
- Clear, colorless liquid
- Formulated using three ingredients known to be active against RNase
RNase Erase™ was formulated using three ingredients known to be active against RNase. It effectively removes high levels of RNase contamination that similar products cannot.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Physical Form:Clear, colorless liquid