Order Entry
Puerto Rico
ContactUsLinkComponent
Finnpipette™ F1 Variable Volume Pipettes, Thermo Scientific
Finnpipette™ F1 Variable Volume Pipettes, Thermo Scientific
Catalog # 490014-006
Finnpipette™ F1 Variable Volume Pipettes, Thermo Scientific
Catalog # 490014-006
Supplier Number:  4641070

Specifications

  • Volume:
    10 - 100 µl
  • Description:
    Finnpipette™ F1 Adjustable Volume Pipettes
  • Compatible with:
    Finntip flex 200, 250 universal, 300, 200 Ext.
  • Cat. No.:
    490014-006
  • Increments:
    0.2 µl
  • Precision:
    1 to 0.2%
  • Color code:
    Yellow
  • Supplier no.:
    4641070

Specifications

About this item

Silver (Ag) ions create an antimicrobial surface that inhibits growth of bacteria, microbes, and fungus.

  • Inhibits bacteria, microbes and fungus
  • Pipetting button enables easy volume adjustment
  • Smooth pipetting action enables better results
  • Soft-touch tip ejection minimizes force needed
  • Limited warranty period

Pipetting button rotates freely when not in locking position.

Simplify volume adjustment with Thermo Scientific™ Finnpipette™ F1 Variable-Volume Pipettes, which include an easy 'set and forget' lock mechanism that features an adjustable finger rest and proprietary soft touch tip ejection. An antimicrobial treatment protects the instrument against contamination.

Finger rest can be adjusted 120° for optimal pipetting position. Light, smooth pipetting action enables better results over longer periods. Super blow-out piston with volumes of 50 µl and below ensures delivery of micro-size drops. Proprietary soft-touch tip ejection minimizes forces needed for tip ejection.