Order Entry
Puerto Rico
ContactUsLinkComponent
1X Phosphate Buffered Saline without Magnesium and Calcium, MP Biomedicals
1X Phosphate Buffered Saline without Magnesium and Calcium, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
1X Phosphate Buffered Saline without Magnesium and Calcium, MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • Sterility:
    Sterile
  • Endotoxin Content:
    <0.15 EU/ml
  • Format:
    Solution
  • pH Range:
    7.0 - 7.6
  • Osmolality:
    270 - 305 mM/kg
  • Formulation:
    Inorganic Salts: Potassium Chloride [KCl]: 200.00 mg/L; Potassium Phosphate Monobasic [KH2PO4]: 200.00 mg/L; Sodium Chloride [NaCl]: 8000.00 mg/L; Sodium Phosphate Dibasic [Na2HPO4]: 1150.00 mg/L

Specifications

Related Information

Frequently Bought Together