ContactUsLinkComponent
Spill-X® Multi-Purpose Treatment Kit, PIG®
Spill-X® Multi-Purpose Treatment Kit, PIG®
Catalog # 89501-394
Supplier:  New Pig
undefined
Spill-X® Multi-Purpose Treatment Kit, PIG®
Catalog # 89501-394
Supplier:  New Pig
Supplier Number:  GEN527
Specifications
  • Description:
    Spill-X® Multi-Purpose Treatment Kit
  • Size:
    6 containers per box
  • Cat. No.:
    89501-394
  • Supplier no.:
    GEN527
Specifications
About this item
Neutralizes smaller spills for safer cleanup and handling.
  • Great for labs
  • Neutralize a wide range of hazardous chemicals and solvents
  • Reduces product waste and disposal costs
Neutralizes smaller spills for safer cleanup and handling – great for labs. Packed with spill control agents that neutralize a wide range of hazardous chemicals and solvents. Granular base lets you use only what you need; reduces product waste and disposal costs. Speeds spill cleanup to limit worker exposure and minimize production downtime.
Ordering information: Kit includes Spill-X-A Acid-Neutralizing Adsorbent, Spill-X-C Caustic-Neutralizing Adsorbent and Spill-X-S Solvent Adsorbent.