Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

6-Fluoro-7-azaindole

1 g
Catalog # 102897-748
Marketsource
Supplier:   Matrix Scientific
 :  113318-1G
6-Fluoro-7-azaindole
Catalog # 102897-748
Supplier:  Matrix Scientific
Marketsource
CAS Number:  898746-42-4
6-Fluoro-7-azaindole

1 g

CAS Number:  898746-42-4
Price
$1,453.85
Log in for availability
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Hazard: Irritant

Synonyms:  6-Fluoro-1H-pyrrolo[2,3-b]pyridine
  • CAS Number:898746-42-4
  • Formula:C₇H₅FN₂
  • MDL Number:MFCD06659668
  • Molecular Weight:136.13 g/mol
Specifications

Specifications

  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.