Order Entry
Puerto Rico
ContactUsLinkComponent
Polyethylene Disposal Bags, PIG®, New Pig
Polyethylene Disposal Bags, PIG®, New Pig
Catalog # 46000-770
Supplier:  New Pig
undefined
Polyethylene Disposal Bags, PIG®, New Pig
Catalog # 46000-770
Supplier:  New Pig
Supplier Number:  BAG202-S

Specifications

  • Capacity:
    10 gal.s
  • Description:
    Polyethylene disposal bags
  • Dimensions:
    18W×30" H
  • Cat. No.:
    46000-770
  • Supplier no.:
    BAG202-S

Specifications

About this item

Tough polyethylene bags resist punctures, tearing and blowouts.

  • Durable thickness
  • Bright yellow bags
  • Resist punctures, tearing and blowouts

Durable 3 mil thickness. Bright yellow bags are boldly labeled 'Hazardous Material - Handle With Care'.