Order Entry
Puerto Rico
ContactUsLinkComponent
Magnesium Standard for AAS, Certipur®, MilliporeSigma
Magnesium Standard for AAS, Certipur®, MilliporeSigma
Catalog # EM1.19788.0100
Supplier:  MilliporeSigma
CAS Number:  
Magnesium Standard for AAS, Certipur®, MilliporeSigma
Catalog # EM1.19788.0100
Supplier:  MilliporeSigma
Supplier Number:  1.19788.0100
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Hazard Class Description:
    Corrosive Materials
  • Cat. No.:
    EM1.19788.0100
  • pH:
    0.5 (H₂O, 20°C)
  • Element:
    Magnesium
  • Supplier no.:
    1.19788.0100

Specifications

About this item

Traceable to SRM from NIST.