Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Bladder Spill Containment Deck, PIG®

Bladder Spill Containment Deck, PIG®

PIG® Bladder Spill Containment Deck, 52" W x 56" L x 5.75" H, 99 gal.
Catalog # 10860-076
Supplier:   New Pig
Supplier #:  PAK530
Bladder Spill Containment Deck, PIG®
Catalog # 10860-076
Supplier:  New Pig
Bladder Spill Containment Deck, PIG®
Bladder Spill Containment Deck, PIG®

PIG® Bladder Spill Containment Deck, 52" W x 56" L x 5.75" H, 99 gal.

Price
$1,144.74
Log in for availability
Each
Product Details & Documents

Patented "flow-through" sump design lets liquid pass freely from one sump to another to help you comply with containment regulations.

  • Exclusive bulkhead fittings
  • Predrilled ports to connect to other decks
  • Decks are ready to use

Modular design features exclusive bulkhead fittings and predrilled ports to connect to other decks; decks are ready to use as soon as they arrive. Integrated bladder box provides 55 gallons of containment beyond deck alone; keeps deck surface at a low 5.75" height.

Specifications
  • Description:
    PIG® Bladder Spill Containment Deck
  • Dimensions:
    52" W x 56" L x 5.75" H
  • Sump capacity:
    99 gal.