ContactUsLinkComponent
SterEM™ Tryptic Soy Agar (TSA), Irradiated
  76200-294
 :  
undefined
SterEM™ Tryptic Soy Agar (TSA), Irradiated
  76200-294
 :  W540
 :  
  • Pack type:
    Plate
  • Description:
    SterEM™ Tryptic soy agar (TSA), irradiated USP, triple bagged
  • Size:
    10/pack
  • Sterility:
    Promotes a higher sterility assurance level of 10⁻⁶
  • Cat. No.:
    76200-294
  • Plate size:
    15×100 mm
  • Application:
    Compounding pharmacy, pharmaceutical manufacturing
Sterile plates for environmental monitoring.
  • New moisture control packaging format prevents moisture buildup and ensures safe and sterile transfer
  • Optically clear polystyrene for distortion free viewing
  • Heavy Duty reduces breakage during shipping
  • Polystyrene petri plates feature raised rings for stacking ability and prevents sliding
  • Sterile by irradiation, validated for Sterility Assurance Level (SAL) of 10⁻⁶
  • Triple bagged, two nylon inner bags and one outer Mylar(R) bag that is heat sealed
Irradiated Tryptic Soy Agar (TSA), USP is recommended for use as a general growth medium for the detection and enumeration of microorganisms. The plates are triple bagged and sterilized by irradiation to promote a higher sterility assurance level.
 : Certified to ISO 13485 - Manufactured in a licensed cGMP facility with the quality management system certified to ISO 13485.