ContactUsLinkComponent
Humidification Chamber, Restek
Humidification Chamber, Restek
Catalog # 10862-928
Supplier:  Restek
undefined
Humidification Chamber, Restek
Catalog # 10862-928
Supplier:  Restek
Supplier Number:  24282
Specifications
  • Description:
    Humidification Chamber
  • Cat. No.:
    10862-928
  • Supplier no.:
    24282
Specifications
About this item
Restek’s humidification chamber is made of acrylic and withstands pressure up to 90 psi.
  • Withstands high pressures
  • Acrylic
  • Easy connection to pressure lines
When cleaning SilcoCan® or TO-Can® canisters, it is important to use humidified air or nitrogen to help remove volatile organic contaminants. Restek’s humidification chamber is made of acrylic and withstands pressure up to 90 psi. The 1/4-inch inlet and outlet compression fittings allow easy connection to pressure lines on your cleaning system. Our humidification chamber also has an easy-to-open lid for filling with water.