Replacement Combination Vacuum/Pressure Gauges, Restek
: Restek
2-inch vacuum/pressure gauges, 316 stainless steel with 1/8" NPT fitting and center back mount.
- Combination gauges
- For use with canisters
Recommended for use with canisters.
2-inch vacuum/pressure gauges, 316 stainless steel with 1/8" NPT fitting and center back mount.
Recommended for use with canisters.