Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Catalog # 490016-842
Supplier: Advansta
Specifications
- Antibody Type:Secondary
- Antigen Symbol:IgG
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Goat
- Reactivity:Human
- Size:500 µL
- Cat. No.:490016-842
- Supplier no.:R-05073-500
Specifications
About this item
No cross-reactivity in immunoblotting applications