About this item
Heparin-binding epidermal growth factor-like growth factor (HB-EGF) is a member of the epidermal growth factor (EGF) family and is expressed by monocytes and macrophages. HB-EGF is the predominant growth factor involved in epithelialization during wound healing. HB-EGF signals through the receptor tyrosine kinase ErbB2 to maintain adult heart homeostasis, and promotes cardiac valve development through binding in high affinity to the epidermal growth factor receptor (EGFR). HB-EGF binds the the ErbB4 receptor tyrosine kinase to mediate implantation of the human blastocyst. HB-EGF also functions as a potent mitogen for fibroblasts and smooth muscle cells.
Animal-free.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: 3T3 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1.0×10⁶ units/mg
- Gene ID:Q99075
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:DT-R|HBEGF|Diphtheria toxin receptor
- Protein/Peptide Name:HB-EGF
- Purity:≥95%
- Molecular Weight:9.9 kDa
- Sequence:MDLQEADLDL LRVTLSSKPQ ALATPNKEEH GKRKKKGKGL GKKRDPCLRK YKDFCIHGEC KYVKELRAPS CICHPGYHGERCHGLSL
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Shipping Temperature:RT
- Tested Applications:Bioassay
- Grade:RUO