Autosampler Preventive Maintenance Kit for Agilent 1100, 1200 HPLC Systems, Restek
Catalog # 10059-396
Supplier: Restek
Specifications
- Description:Autosampler Preventive Maintenance Kit for Agilent 1100/1200
- Cat. No.:10059-396
- Model:1100, 1200
- Includes:Rotor seal, piston seals (2), needle assembly, needle seat, finger caps (3)
- Supplier no.:25271