ContactUsLinkComponent
Ternary Tee Mixer, Restek
Ternary Tee Mixer, Restek
Catalog # 10853-382
Supplier:  Restek
undefined
Ternary Tee Mixer, Restek
Catalog # 10853-382
Supplier:  Restek
Supplier Number:  25138
Specifications
  • Volume:
    25 µl
  • Description:
    HyperShear Static Mixer, Ternary Tee Mixer 25uL - Stainless Steel
  • Cat. No.:
    10853-382
  • Supplier no.:
    25138
Specifications
About this item
An efficient cross-flow shearing mechanism and interchangeable cartridges produce vortex shear mixing over a wide range of flows.
  • Reduced baseline noise
  • Increased sensitivity
  • Improved gradient accuracy for more reproducible results
  • Increased reaction efficiency in post-column derivatization
Stainless steel.