ContactUsLinkComponent
LC Piston Seal Insertion Tool, Restek
LC Piston Seal Insertion Tool, Restek
Catalog # 10853-846
Supplier:  Restek
undefined
LC Piston Seal Insertion Tool, Restek
Catalog # 10853-846
Supplier:  Restek
Supplier Number:  21356
Specifications
  • Description:
    LC Piston Seal Insertion Tool
  • Cat. No.:
    10853-846
  • Supplier no.:
    21356
Specifications
About this item
Simplify pump maintenance.
Use one end to remove your old seal, then simply slip your new seal on the other end and push it flush into position. The tool cannot mar the surrounding metal surface of the pump housing.