About this item
Oncostatin M (OSM) is a cytokine that is produced by macrophages, dendritic cells, and T lymphocytes during inflammatory events. The Type-I and Type-II OSM receptors are located on the cell surface of endothelial and tumor cells, contain the glycoprotein 130 (gp130) subunit, and activate the JAK/STAT signaling pathway. OSM functions to inhibit tumor cell proliferation, induce liver stem cell maturation, regulate cytokine production during hematopoiesis and inflammation, stimulate bone formation, and promote nervous system development.
- Animal-free
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤1
- Biological Activity:Bioactivity assay: TF-1 cell proliferation, ED50 ≤4 ng/ml, Bioactivity ≥2.5×10⁵ units/mg
- Gene ID:P13725
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Onc-M
- Protein/Peptide Name:Oncostatin M
- Purity:≥95%
- Molecular Weight:23.8 kDa
- Sequence:MAAIGSCSKE YRVLLGQLQK QTDLMQDTSR LLDPYIRIQG LDVPKLREHC RERPGAFPSE ETLRGLGRRG FLQTLNATLGCVLHRLADLE QRLPKAQDLE RSGLNIEDLE KLQMARPNIL GLRNNIYCMA QLLDNSDTAE PTKAGRGASQ PPTPTPASDA FQRKLEGCRFLHGYHRFMHS VGRVFSKWGE SPNRSRRHSP HQALRKGVRR
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay